Learn More
Potassium (cyanomethyl)trifluoroborate, 95%, ACROS Organics™
Brand: Acros Organics 444860050
Code : 29
Additional Details : CAS Number : 888711-58-8 Weight : 0.05500kg Transport : UN number : 3439 Chem class : 6.1 Pack group : III
Quantity | 5g |
---|
Chemical Identifiers
888711-58-8 | |
146.95 | |
potassium cyanomethyl trifluoroborate, c2h2bf3n.k, potassium cyanomethyl trifluoroboranuide, potassium cyanomethyl trifluoro borate 1- |
C2H2BF3KN | |
IJENQMHWPRCCBA-UHFFFAOYSA-N | |
23666193 |
Specifications
Potassium (cyanomethyl)trifluoroborate | |
5g | |
95% | |
potassium cyanomethyl trifluoroborate, c2h2bf3n.k, potassium cyanomethyl trifluoroboranuide, potassium cyanomethyl trifluoro borate 1- | |
146.95 | |
146.95 |
151°C to 156°C | |
888711-58-8 | |
C2H2BF3KN | |
IJENQMHWPRCCBA-UHFFFAOYSA-N | |
23666193 | |
95% |
Safety and Handling
- Potassium (cyanomethyl)trifluoroborate
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.