Learn More
Potassium Dichromate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™
High quality chemicals for laboratory and specialized industrial use
Brand: JT Baker 3090-01
Additional Details : CAS Number : 7778-50-9 Weight : 0.64000kg Transport : UN number : 3288 Chem class : 6.1 Pack group : III
Packaging | Poly Bottle |
---|---|
Quantity | 500g |
Chemical Identifiers
7778-50-9 | |
294.182 | |
potassium dichromate, potassium bichromate, kaliumdichromat, iopezite, dipotassium bichromate, dipotassium dichromate, potassium dichromate vi, dichromic acid dipotassium salt, dipotassium dichromium heptaoxide, bichromate of potash | |
CHEBI:53444 | |
[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+] |
Cr2K2O7 | |
KMUONIBRACKNSN-UHFFFAOYSA-N | |
24502 | |
dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium |
Specifications
Potassium Dichromate | |
Cr2K2O7 | |
KMUONIBRACKNSN-UHFFFAOYSA-N | |
dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium | |
24502 | |
294.19 | |
BAKER ANALYZED™ ACS Reagent | |
1L = 2.68kg |
7778-50-9 | |
potassium dichromate, potassium bichromate, kaliumdichromat, iopezite, dipotassium bichromate, dipotassium dichromate, potassium dichromate vi, dichromic acid dipotassium salt, dipotassium dichromium heptaoxide, bichromate of potash | |
[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+] | |
294.182 | |
CHEBI:53444 | |
Crystals | |
Poly Bottle | |
500g |