missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Dihydrogen Orthophosphate, Certified AR for Analysis, Fisher Chemical™
27.65 EUR valid until 2024-12-31
Use promo code "22914" to get your promotional price.
28.55 EUR valid until 2024-04-30
Use promo code "21741" to get your promotional price.
Use promo code "22914" to get your promotional price.
28.55 EUR valid until 2024-04-30
Use promo code "21741" to get your promotional price.
Click to view available options
Quantity:
250 g
500 g
1 kg
3 kg
Description
Potassium Dihydrogen Orthophosphate, Certified AR for Analysis, Fisher Chemical, Quantity: 500g, CAS: 7778-77-0, White, Melting Point: 253 deg.C, Molecular Formula: H2KO4P, Molecular Weight: 136.084, Packaging: Plastic powder jar, pH: 4.5
Specifications
Melting Point | 253°C |
Color | White |
pH | 4.5 |
Physical Form | Solid |
Quantity | 250 g |
IUPAC Name | potassium;dihydrogen phosphate |
Formula Weight | 136.09g/mol |
Packaging | Plastic powder jar |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'