missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium hexachloroplatinate(IV), ca. 40% Pt, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 250.0°C |
Color | Black |
Physical Form | Powder |
Quantity | 25 g |
Linear Formula | K2PtCl6 |
Merck Index | 15,7757 |
Solubility Information | Solubility in water: 50g/L (95°C). Other solubilities: insoluble in alcohol |
IUPAC Name | dipotassium;hexachloroplatinum(2-) |
Formula Weight | 486.01 |
Percent Purity | ∼40% (Pt) |
Show More |
Safety and Handling
Product Identifier
- Potassium hexachloroplatinate(IV)
- Danger
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H301-Toxic if swallowed.
- H290-May be corrosive to metals.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'