missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Iodate, Certified AR for Analysis, Fisher Chemical™
40.65 EUR valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
100 g
250 g
500 g
1 kg
Description
Potassium Iodate, Certified AR for Analysis
Specifications
Melting Point | 560°C |
Color | White |
Physical Form | Powder/Solid |
Quantity | 100 g |
Formula Weight | 214g/mol |
Packaging | Plastic powder jar |
Safety and Handling
Product Identifier
- Potassium iodate
- Danger
- Oxidising solid Category 2
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H272-May intensify fire; oxidiser.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'