missing translation for 'onlineSavingsMsg'
Learn More

Potassium L-tartrate hemihydrate, 99%, Thermo Scientific Chemicals

Catalog Number p-7021991
Click to view available options
Quantity:
250 g
1000 g
5000 g
This item is not returnable. View return policy
6100-19-2
C8H14K4O13
474.58
MFCD00150136,MFCD00150136
DFVWEDQSAWCYPI-UHFFFAOYNA-N
potassium tartrate dibasic hemihydrate
131855961
O.[K].[K].[K].[K].OC(C(O)C(O)=O)C(O)=O.OC(C(O)C(O)=O)C(O)=O
This item is not returnable. View return policy
6100-19-2
C8H14K4O13
474.58
MFCD00150136,MFCD00150136
DFVWEDQSAWCYPI-UHFFFAOYNA-N
potassium tartrate dibasic hemihydrate
131855961
O.[K].[K].[K].[K].OC(C(O)C(O)=O)C(O)=O.OC(C(O)C(O)=O)C(O)=O

Potassium L-tartrate hemihydrate is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium L-tartrate hemihydrate is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Soluble in water.

Notes
Store in cool, dry conditions in a well sealed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 155°C (in -H2O)
Density 1.98
Boiling Point 200°C to 220°C
Quantity 250 g
Beilstein 6119985
Merck Index 14,7677
Solubility Information Soluble in water.
IUPAC Name (2R,3R)-2,3-dihydroxybutanedioic acid;potassium;hydrate
Formula Weight 235.29 (226.27 Anhydrous)
Percent Purity 99%
Chemical Name or Material Potassium L-tartrate hemihydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'