Learn More
Thermo Scientific™ Potassium nitrate, 99+%, for biochemistry
Chemical Identifiers
| CAS | 7757-79-1 |
|---|---|
| Molecular Formula | KNO3 |
| Molecular Weight (g/mol) | 101.1 |
| InChI Key | FGIUAXJPYTZDNR-UHFFFAOYSA-N |
| Synonym | potassium nitrate, saltpeter, nitre, nitrate of potash, vicknite, nitric acid potassium salt, nitric acid, potassium salt, saltpetre, kaliumnitrat, kalii nitras |
| PubChem CID | 24434 |
| ChEBI | CHEBI:63043 |
| IUPAC Name | potassium;nitrate |
| SMILES | [N+](=O)([O-])[O-].[K+] |
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 7757-79-1 | |
| 101.1 | |
| potassium nitrate, saltpeter, nitre, nitrate of potash, vicknite, nitric acid potassium salt, nitric acid, potassium salt, saltpetre, kaliumnitrat, kalii nitras | |
| CHEBI:63043 | |
| [N+](=O)([O-])[O-].[K+] |
| KNO3 | |
| FGIUAXJPYTZDNR-UHFFFAOYSA-N | |
| 24434 | |
| potassium;nitrate |
Specifications
| 7757-79-1 | |
| KNO3 | |
| 15,7769 | |
| Solubility in water: 320g/L (20°C). Other solubilities: soluble in glycerol,slightly soluble in alcohol | |
| [N+](=O)([O-])[O-].[K+] | |
| 101.1 | |
| CHEBI:63043 | |
| 99+% | |
| Potassium nitrate |
| 99+% | |
| KNO3 | |
| potassium nitrate, saltpeter, nitre, nitrate of potash, vicknite, nitric acid potassium salt, nitric acid, potassium salt, saltpetre, kaliumnitrat, kalii nitras | |
| FGIUAXJPYTZDNR-UHFFFAOYSA-N | |
| potassium;nitrate | |
| 24434 | |
| 101.1 | |
| Biochemistry |
Safety and Handling
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 231-818-8
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.