missing translation for 'onlineSavingsMsg'
Learn More

Potassium periodate, 99.5%, extra pure, ACROS Organics™

Brand:  Acros Organics 198391000

Code : 29

Additional Details : CAS Number : 7790-21-8 Weight : 0.15000kg Transport : UN number : 1479 Chem class : 5.1 Pack group : II


Packaging Plastic bottle
Quantity 100g

 View more versions of this product

Product Code. 10022510

  • 70.92 € / 100g

In stock, ready to ship: 9

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Oxidation reagent
Specifications

Chemical Identifiers

7790-21-8
230
potassium periodate, potassium metaperiodate, unii-q3loc6p66l, q3loc6p66l, periodic acid hio4 , potassium salt, potassium ion io4, ssmicroaea 1/4 o, acmc-20ajue, ssmicroaea 1/4 oni, periodic acid hio4 , potassium salt 1:1
potassium;periodate
IKO4
FJVZDOGVDJCCCR-UHFFFAOYSA-M
516896
[O-]I(=O)(=O)=O.[K+]

Specifications

Potassium periodate
99.4% min. (Iodometry)
KIO4
15,7775
Solubility in water: 0.42 g/100mL (20°C).
[O-]I(=O)(=O)=O.[K+]
230
230
99.5%
1ppm max.
Plastic bottle
White
100g
7790-21-8
IKO4
01,809; 02,311; 04,373; 05,507; 06,455
potassium periodate, potassium metaperiodate, unii-q3loc6p66l, q3loc6p66l, periodic acid hio4 , potassium salt, potassium ion io4, ssmicroaea 1/4 o, acmc-20ajue, ssmicroaea 1/4 oni, periodic acid hio4 , potassium salt 1:1
FJVZDOGVDJCCCR-UHFFFAOYSA-M
potassium;periodate
516896
Crystalline Powder
Extra Pure
0.01% max.
Solubility in water: 0.42 g/100mL (20°C).
582.0°C
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Potassium periodate

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H272-May intensify fire; oxidiser.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS Documents