missing translation for 'onlineSavingsMsg'
Learn More

Potassium sulfate, pure, Thermo Scientific Chemicals

Catalog Number 11954241
Click to view available options
Quantity:
1 kg
5 kg
25 kg
Packaging:
Fibre drum
Plastic bottle
7778-80-5
K2O4S
174.25
MFCD00011388
OTYBMLCTZGSZBG-UHFFFAOYSA-L
potassium sulfate, dipotassium sulfate, potassium sulphate, sulfuric acid dipotassium salt, sal polychrestum, sulfuric acid, potassium salt, arcanum duplicatum, kalium sulphuricum, caswell no. 702, sulfuric acid, dipotassium salt
24507
CHEBI:32036
[K+].[K+].[O-]S([O-])(=O)=O
7778-80-5
K2O4S
174.25
MFCD00011388
OTYBMLCTZGSZBG-UHFFFAOYSA-L
potassium sulfate, dipotassium sulfate, potassium sulphate, sulfuric acid dipotassium salt, sal polychrestum, sulfuric acid, potassium salt, arcanum duplicatum, kalium sulphuricum, caswell no. 702, sulfuric acid, dipotassium salt
24507
CHEBI:32036
[K+].[K+].[O-]S([O-])(=O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 1689.0°C
Melting Point 1067.0°C
Color White
pH 5.5 to 8.5
Physical Form Crystalline Powder
Quantity 1 kg
Linear Formula K2SO4
Merck Index 15,7793
Solubility Information Solubility in water: 110g/L (20°C). Other solubilities: insoluble in alcohol
IUPAC Name dipotassium sulfate
Formula Weight 174.25
Percent Purity ≥99.0%
Grade Pure
Loss on Drying 1% max. (130°C)
Packaging Plastic bottle
Chemical Name or Material Potassium sulfate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'