missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Potassium tantalum isopropoxide, 2.5% w/v in isopropanol

Brand:  Alfa Aesar™ 045651.AK

Code : NEW

Additional Details : CAS Number : 21864-26-6 Weight : 0.00025kg Transport : UN number : 1219 Chem class : 1 Pack group : II

Quantity 250mL

 View more versions of this product

Product Code. 15417045

  • 161.92 € / 250mL


Estimated Shipment date
from Supplier 26-02-2018
Add to basket

Specifications

Chemical Identifiers

Potassium tantalum isopropoxide
C18H42KO6Ta
574.57
UN1219
21864-26-6
KTa[OCH(CH3)2]6
MFCD00210644

Specifications

Potassium tantalum isopropoxide
Liquid
KTa[OCH(CH3)2]6
MFCD00210644
Alcohol-like
2.5% w/v in isopropanol
12°C (54°F)
21864-26-6
C18H42KO6Ta
574.57
UN1219
Colorless to pale yellow
250mL
Moisture Sensitive
Videos
SDS