missing translation for 'onlineSavingsMsg'
Learn More

Potassium tert-butoxide, pure, 1M solution in THF, AcroSeal™, Thermo Scientific Chemicals

Brand:  Thermo Scientific Acros 371220025

Code : 29

Additional Details : CAS Number : 865-47-4 Weight : 2.25500kg Transport : UN number : 2920 Chem class : 8 Pack group : I


 View more versions of this product

Product Code. 11975851

  • 2103.00 € / 2.50 L
Estimated Shipment: 06-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Sea freight only: For EU stock allow up to 15 days delivery.

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Potassium tert-butoxide
Colorless to Orange-Yellow
2.5 L
(CH3)3COK
−19°C
LPNYRYFBWFDTMA-UHFFFAOYSA-N
potassium;2-methylpropan-2-olate
23665647
Pure
AcroSeal™ Glass Bottle
0.9020g/mL
865-47-4,109-99-9
Solution
C4H9KO
01,911; 02,336; 03,233; 04,399; 05,544; 06,477; 08,407; 09,380; 10,323; 11,432; 12,97; 14,264; 17,289
potassium tert-butoxide, potassium tert-butanolate, potassium t-butoxide, potassium 2-methylpropan-2-olate, potassium tert-butylate, kotbu, 2-methyl-2-propanol, potassium salt, tert-butoxypotassium, potassium-t-butoxide, t-buok
CC(C)(C)[O-].[K+]
112.213
112.21
0.95 to 1.10M
0.902
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Potassium tert-butoxide, 1M solution in tetrahydrofuran

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity Category 3

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only