missing translation for 'onlineSavingsMsg'
Learn More

Potassium thiocyanate, 99+%, for analysis, ACROS Organics™

Brand:  Acros Organics 196585000

Code : 29

Additional Details : CAS Number : 333-20-0 Weight : 0.55000kg


Packaging Plastic bottle
Quantity 500g

 View more versions of this product

Product Code. 10002490

  • 51.13 € / 500g

In stock, ready to ship: 29

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Indicator
Specifications

Chemical Identifiers

333-20-0
97.18
ZNNZYHKDIALBAK-UHFFFAOYSA-M
516872
potassium;thiocyanate
CKNS
MFCD00011413
potassium thiocyanate, potassium rhodanate, potassium rhodanide, rodanca, potassium sulfocyanate, rhodanide, rhocya, potassium thiocyanide, kscn, thiocyanic acid, potassium salt
CHEBI:30951
C(#N)[S-].[K+]

Specifications

Potassium thiocyanate
333-20-0
CKNS
MFCD00011413
01,954
potassium thiocyanate, potassium rhodanate, potassium rhodanide, rodanca, potassium sulfocyanate, rhodanide, rhocya, potassium thiocyanide, kscn, thiocyanic acid, potassium salt
ZNNZYHKDIALBAK-UHFFFAOYSA-M
potassium;thiocyanate
516872
97.18
99+%
50ppm max
1.8860g/mL
30mg I2/100g max.
Plastic bottle
Solubility in water: 2170g/L (20°C). Other solubilities: soluble in alcohol and acetone
50ppm max.
170.0°C to 179.0°C
500g
p.a.
99% min. (Argentometry)
KSCN
10ppm max.
15, 7807
Solubility in water: 2170g/L (20°C). Other solubilities: soluble in alcohol and acetone
C(#N)[S-].[K+]
97.18
CHEBI:30951
Crystalline Powder
Analysis
50ppm max.
2ppm max.
2ppm max.
200ppm max.
1.886
White
5.3 to 8.7 (5% soln. at 20°C)
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Potassium thiocyanate

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3

Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
  • EUH032-Contact with acids liberates very toxic gas.

Precautionary Statement
  • P233-Keep container tightly closed.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

SDS Documents