Learn More
Bel-Art™ SP Scienceware™ Poxygrid Steel Test Tube Racks
Chemical and corrosion resistant steel test tube racks in different colors for easy identification.
Brand: Bel-Art™ SP Scienceware™ F18767-0000
Code : NEW
Additional Details : Weight : 0.32380kg
Description
Chemical and corrosion resistant steel test tube racks in different colors for easy identification.
- Chemical and corrosion-resistant steel wire racks are coated with heat-cured epoxy
- Orange racks can be used for biohazard warning, yellow for radioactive warning
- Epoxy coatings are unaffected by organic solvents, salts, weak acids and weak alkalis
- Will withstand a temperature range of −223°C (−369°F) to 121°C (250°F)
- All racks have square holes, three decks, a slotted grid bottom and reinforced edges
Specifications
Test Tube Rack | |
Autoclavable | |
Tubes (20 to 25 mm) | |
40 | |
Non-sterile | |
-223°C to +121°C | |
25 mm |
4 x 10 | |
Green | |
Epoxy Coated Steel | |
Rectangle | |
20 - 25mm Tubes | |
26.3 x 11.6 x 8.3 cm (10.38 x 4.56 x 3.25 in.) |