missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pretreatment Cartridges and Filters

Product Code. 11387101 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Filter
Filter with Stabilizer
Prefilter
Prefilter MQ Cartridge
Pretreatment Cartridge
Pretreatment Filter
Stabilizing Cartridge
Unit Size:
1 unit
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type unitSize
11387101 Stabilizing Cartridge Each
11327151 Pretreatment Cartridge Each
11766259 Prefilter MQ Cartridge Each
15322178 Prefilter 1 unit
12756208 Filter with Stabilizer Each
11367151 Pretreatment Filter Each
12818111 Filter Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 11387101 Supplier Thermo Scientific™ Supplier No. 06.5452
  • ONLINE EXCLUSIVE PRICE
    169.00 €
    163.00 € / Pack of 1
    Save 6.00 €
    3% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Choose one of these Thermo Scientific™ Pretreatment Cartridges and Filters when the quality of your feed water doesn't match the requirements for your point of use water purification system.

Use Thermo Scientific™ Pretreatment Cartridges and Filters when the quality of your feed water doesn't match the requirements for your Thermo Scientific™ Barnstead™ water purification system.

Use pretreatment when your feed water quality doesn't match the requirements for your system.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) LabTower TII, Pacific RO, Pacific TII
Type Stabilizing Cartridge
Includes Hardness Stabilizing Cartridge
Length (English) 10 ft.
Height (English) 10 ft.
Height (Metric) 25.4 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.