Learn More
Invitrogen™ PRIMER, 5'ALEXA 660 200 NMOL
Custom Primer
Brand:  Invitrogen™ 10660014
Additional Details : Weight : 0.45000kg
Product Code. 10052472
  • 248.00 € / Each
Estimated Shipment: 30-04-2024
to see stock.
Description
Description
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10μmole. Select desired purification method (normal or reversed-phase chromatography, HPLC, or PAGE).
Specifications
Specifications
5' Alexa 660 Primer, 200 nmol
5' Alexa 660 Primer
5' Labeled
200 nmol