Learn More
Invitrogen™ PRIMER 5' FLO 1 UMOL
Custom Primer
Brand: Invitrogen™ 10918019
193.34 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Additional Details : Weight : 0.00100kg
Description
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10μmole. Select desired purification method (normal or reversed-phase chromatography, HPLC, or PAGE).Specifications
5' FLO Primer, 1μmol | |
5' FLO Primer | |
5' Labeled | |
1 μmol |