Learn More
Invitrogen™ PRIMER 5' HEX 1 UMOL
Custom Primer
Brand:  Invitrogen™ 11862042
131.33 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Additional Details : Weight : 0.45000kg
Product Code. 10550333
  • 197.00 € / Each
Estimated Shipment: 19-04-2024
to see stock.
Description
Description
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10μmole. Select desired purification method (normal or reversed-phase chromatography, HPLC, or PAGE).
Specifications
Specifications
5' HEX Primer, 1μmol
5' HEX Primer
5' Labeled
1 μmol