Learn More
Invitrogen™ PRIMER INOSINE
Custom Primer
Brand: Invitrogen™ 10624013
Additional Details : Weight : 0.00100kg
Description
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10μmole. Select desired purification method (normal or reversed-phase chromatography, HPLC, or PAGE).Specifications
Inosine Primer, 50 nmol | |
Inosine Primer | |
50 nmol |