Learn More
Chemsplash Pro+4 Type 456 Coverall Taped
Description
Chemsplash Pro +4 Disposable Coverall
- 67GSM Microporous Laminate Fabric
- Fully Taped Seams
- Three Piece Hood
- Elasticated Cuffs with Thumb Loops
- Elasticated Hood, Back & Ankles
- Two Way Zip with Zip Flap
- Silicone and Latex Free, PFAS Free
- Conforms to ISO 14644-1:2015 Cleanroom 5 - 9
- Low Linting
- Anti-Static
Description: Chemsplash Pro +4 Disposable Coverall is made from a 67GSM microporous laminated Cat III Type 4B/5B/6B fabric. All seams are taped offering a stronger and more effective barrier to liquids and particulate matter. Chemsplash Pro +4 fabric is both anti-static to EN1149-5:2008 and is resistant to infectious agents under EN14126, complying with resistance to viral and bacterial infection. Making it an ideal choice for Pharmaceutical, Cleanroom, Semi-Conductor manufacturing and Infection Contamination Control. Conforms to ISO 14644-1:2015 Cleanroom 5 - 9.
Specifications
Specifications
| Product Type | Cleanroom Coveralls |
| Size | Large |
| Gender | Unisex |
| Material | Microporous Composite Fabric |
| Color | White |
| Disposable | Single-use |
| Waist Style | Elastic |
| Ankle Style | Elastic |
| Cuff Style | Elastic, Thumb Loop |
| Neck Style | Hooded |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.