missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ProcartaPlex™ Human Viral Reagent Kit

Product Code. 17158875
Click to view available options
Quantity:
96 Tests
Unit Size:
Pack of 96
This item is not returnable. View return policy

Product Code. 17158875

Brand: Invitrogen™ EPX01010430901

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Designed to be used with the Coronavirus Simplex kits to combine single analytes into a multiplex assay

  • The ProcartaPlex Viral Reagent Kit is designed to be used with the Coronavirus Simplex kits to combine single analytes into a multiplex assay.
  • This provides maximum flexibility to design assay panels as needed. This kit includes all reagents except for the target-specific beads (sold separately) to run the assay.
  • ProcartaPlex immunoassays are an ideal solution for assessing multiple protein biomarkers in a single sample.
  • Suitable sample types: serum, plasma
  • Sample volume: serum, plasma: 25 μL
  • Reported application: multiplex immunoassay
TRUSTED_SUSTAINABILITY

Specifications

Product Type Basic and Setup Kits
Quantity 96 Tests
Species Human
Combinability Combinable
Assay Sensitivity See Certificate of Analysis
For Use With (Equipment) Luminex Instruments
Product Line ProcartaPlex™
Content And Storage

• 1 vial Biotinylated Detection Antibody Mix (50X)
• 2 vials Human Control-High
• 2 vials Human Control-Medium
• 2 vials Human Control-Low
• 1 bottle Reading Buffer (1X)
• 1 bottle Wash Buffer (10X)
• 1 bottle Assay Buffer (1X)
• 1 bottle Detection Antibody Diluent (1X)
• 8-tube strip
• Adhesive film
• Flat bottom 96-well plate, black

Store at 2°C to 8°C.

Shipping Condition Wet Ice

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.