missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GAGE2D Protein
A cDNA sequence encoding the GAGE2D was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11925-1mg
Specifications
GAGE2D Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSWRGRS TYRPRPRRYV EPPEMIGPMR PEQFSDEVEP ATPEEGEPAT QRQDPAAAQE GEDEGASAGQ GPKPEADSQE QGHPQTGCEC EDGPDGQEMD PPNPEEVKTP EEGEKQSQC | |
Greater than 95.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
GAGE2D protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.1M NaCl. |