missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse LAMP2 Protein
A cDNA sequence encoding the LAMP2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12531-1mg
Specifications
LAMP2 Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
LIVNLTDSKG TCLYAEWEMN FTITYETTNQ TNKTITIAVP DKATHDGSSC GDDRNSAKIM IQFGFAVSWA VNFTKEASHY SIHDIVLSYN TSDSTVFPGA VAKGVHTVKN PENFKVPLDV IFKCNSVLTY NLTPVVQKYW GIHLQAFVQN GTVSKNEQVC EEDQTPTTVA PIIHTTAPST TTTLTPTSTP TPTPTPTPTV GNYSIRNGNT TCLLATMGLQ LNITEEKVPF IFNINPATTN FTGSCQPQSA QLRLNNSQIK YLDFIFAVKN EKRFYLKEVN VYMYLANGSA FNISNKNLSF WDAPLGSSYM CNKEQVLSVS RAFQINTFNL KVQPFNVTKG QYSTAQDCSA DEDNLEHHHH HH | |
Greater than 95.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Mouse | |
His | |
LAMP2 protein solution (0.5 mg/ml) containing Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |