Learn More
Merck Millipore Proguard™ TS2 Pretreatment Pack
For optimal protection of reverse osmosis membranes. Proguard™ TS2 Pretreatment Packs are designed to remove large particles and organic compounds, softening water, and removing chlorine from tap water.
Brand: Merck Millipore PR0G0T0S2
Additional Details : Weight : 1.66500kg
Description
For optimal protection of reverse osmosis membranes. Proguard™ TS2 Pretreatment Packs are designed to remove large particles and organic compounds, softening water, and removing chlorine from tap water.
- Activated carbon
- Anti-scaling
- Depth filtration
Specifications
Activated Carbon, Anti-Scaling, Depth Filtration | |
Optimal Protection of Reverse Osmosis Membranes (with Bacteriostatic Activated Carbon) | |
1 Each | |
Pretreatment Pack |