Learn More
Promega CellTiter-Glo™ Luminescent Cell Viability Assay Kit
The most sensitive, fastest, homogeneous cell viability assay. Based on luminescent detection of ATP.
POA
Specifications
For Use With (Application) | Automated High Throughput Screening, Cell Proliferation Assays, Cytotoxicity Assays, Determining the number of viable cells in culture based on the amount of ATP present |
---|---|
For Use With (Equipment) | 96- or 384-well Plates |
Includes: CellTiter-Glo Buffer; CellTiter-Glo Substrate (lyophilized)
Description
- A Luminescent Assay to Determine the Number of Viable Cells in Culture
- A homogeneous add-mix-measure format reduces the number of steps
- Higher level of detection requires fewer cells per assay
- Process plates consecutively with a very stable luminescent signal
U.S. Pat. Nos. 6,602,677 and 7,241,584 and other patents and patents pending.
U.S. Pat. No. 7,770,310 and other patents and patents pending.
U.S. Pat. Nos. 7,083,911, 7,452,663 and 7,732,128 and other patents and patents pending.
U.S. Pat. No. 7,741,067 and other patents and patents pending.
The method of recombinant expression of Coleoptera luciferase is covered by U.S. Patents 5,583,024; 5,674,713; and 5,700,673.
Specifications
Automated High Throughput Screening, Cell Proliferation Assays, Cytotoxicity Assays, Determining the number of viable cells in culture based on the amount of ATP present | |
96- or 384-well Plates |