missing translation for 'onlineSavingsMsg'
Learn More

Propidium iodide, 1mg/ml aqueous soln., Thermo Scientific Chemicals

Catalog Number 15417999
Click to view available options
Quantity:
10 mL
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price

 

Membrane impermeant, phenanthridinium intercalator dye excited by mercury or xenon-arc lamps or argon-ion laser. Selectively stains dead cells and a counter stain for multicolor fluorescent images.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Membrane impermeant, phenanthridinium intercalator dye excited by mercury or xenon-arc lamps or argon-ion laser. Selectively stains dead cells and a counter stain for multicolor fluorescent images.

Solubility
Diffic
TRUSTED_SUSTAINABILITY
CAS 25535-16-4
Color Orange-Red
Molecular Formula C27H34I2N4
MDL Number MFCD00011921
Quantity 10 mL
Beilstein 3843838
Sensitivity Light sensitive
Synonym propidium iodide, propidium diiodide, 3,8-diamino-5-3-diethyl methyl ammonio propyl-6-phenylphenanthridin-5-ium iodide, unii-tp416o228t, 3,8-diamino-5-3-diethylmethylammonio propyl-6-phenylphenanthridinium diiodide, 3,8-diamino-5-3-diethylaminopropyl-6-phenylphenanthridinium iodide methiodide, phenanthridinium, 3,8-diamino-5-3-diethylmethylammonio propyl-6-phenyl-, diiodide
Solubility Information Difficult to mix.
InChI Key XJMOSONTPMZWPB-UHFFFAOYSA-M
SMILES CC[N+](C)(CC)CCC[N+]1=C2C=C(C=CC2=C3C=CC(=CC3=C1C4=CC=CC=C4)N)N.[I-].[I-]
IUPAC Name 3-(3,8-diamino-6-phenylphenanthridin-5-ium-5-yl)propyl-diethyl-methylazanium;diiodide
Molecular Weight (g/mol) 668.406
PubChem CID 104981
ChEBI CHEBI:51240
Physical Form Liquid
Chemical Name or Material Propidium iodide, 1mg/mL aqueous soln.
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'