missing translation for 'onlineSavingsMsg'
Learn More

Propyl isothiocyanate, 98%, ACROS Organics™

Brand:  Acros Organics 305310250

Code : 29

Additional Details : CAS Number : 628-30-8 Weight : 0.07500kg Transport : UN number : 2924 Chem class : 3 Pack group : II

Packaging Glass bottle
Quantity 25g

 View more versions of this product

Product Code. 10003740

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 01 885 5854  for assistance.
View alternative products »


Specifications

Chemical Identifiers

628-30-8
101.167
KKASGUHLXWAKEZ-UHFFFAOYSA-N
69403
CCCN=C=S
C4H7NS
MFCD00004823
propyl isothiocyanate, propylisothiocyanate, propane, 1-isothiocyanato, n-propyl isothiocyanate, isothiocyanic acid propyl ester, isothiocyanic acid, propyl ester, isothiocyanic acid n-propyl ester, 1-propyl isothiocyanate, propanisothiocyanate, 1-isothiocyanatopropan
1-isothiocyanatopropane

Specifications

Propyl isothiocyanate
Authentic
1.5030 to 1.5050 (20°C, 589nm)
25g
628-30-8
C4H7NS
MFCD00004823
KKASGUHLXWAKEZ-UHFFFAOYSA-N
1-isothiocyanatopropane
69403
Liquid
0.9780g/mL
Glass bottle
Yellow
98%
98%
CH3CH2CH2NCS
propyl isothiocyanate, propylisothiocyanate, propane, 1-isothiocyanato, n-propyl isothiocyanate, isothiocyanic acid propyl ester, isothiocyanic acid, propyl ester, isothiocyanic acid n-propyl ester, 1-propyl isothiocyanate, propanisothiocyanate, 1-isothiocyanatopropan
CCCN=C=S
101.167
101.17
≥97.5%
Videos
SDS