missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Protease-Free Bovine Albumin, Thermo Scientific Chemicals
Derived from bovine blood.
115.00 € - 765.00 €
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15855068
|
Thermo Scientific
J10867.09 |
10 g |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15875068
|
Thermo Scientific
J10867.22 |
100 g |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15865068
|
Thermo Scientific
J10867.18 |
50 g |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Purity grade: Molecular Biology Grade
- Assay (electrophoresis): ≥98.0%
- Protein: ≥96.0%
- MDLNumber: MFCD00130384
- CAS: 9048-46-8
RUO – Research Use Only