Learn More
Fisher BioReagents™ Protease Inhibitor Cocktail I
Reconstitute with 1mL of water to produce a 100X concentrate solution. Fisher Bioreagents Protease Inhibitor Cocktail I is a lyophilized blend of five inhibitors for inhibiting a broad range of proteases and esterases.
Brand: Fisher BioReagents™ 12801640
Code : 32
Additional Details : Weight : 0.10000kg
Description
- Specificity: Serine Proteases, Esteroses, Cysteine Proteases, Metalloproteases and Trypsin like Proteases
Specifications
Protease Inhibitor Cocktail | |
Store at -20°C | |
1 Vial | |
AEBSF E-64 Leupeptin, Aprotinin EDTA |
Safety and Handling
- Protease Inhibitor Cocktail I (General)
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash hands thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : EDTA Disodium, AEBSFHCl, Aprotinin