missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ Protease Inhibitor Cocktail III for Mammalian Cells

Makes 1mL of 100X concentrate. Fisher Bioreagents Protease Inhibitor Cocktail III for Mammalian Cells is a liquid blend of protease inhibitors optimized for mammalian cells.

Brand:  Fisher BioReagents™ 12831640

Code : 32

Additional Details : Weight : 0.00900kg


Product Code. 12831640

  • 217.30 € / 1mL
Estimated Shipment: 29-07-2024
to see stock.
Add to basket

Description

Description

  • Contains: AEBSF HCl, Aprotinin, E-64, Bestatin, Leupeptin and Pepstatin
Specifications

Specifications

Store at -20°C
Liquid
1 mL
AEBSF HCl, Aprotinin, E-64, Bestatin, Leupeptin and Pepstatin
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Protease Inhibitor Cocktail III

Signal Word
  • Warning

Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : AEBSF HCl, Bestatin,Leupeptin, Pepstatin A, E-64, Aprotinin

SDS
Documents

Documents

Certificates
Special Offers

Special Offers