Learn More
Fisher BioReagents™ Protease Inhibitor Cocktail III, Animal Free (For Mammalian Cells)
Supplied in 1mL DMSO. Fisher Bioreagents Protease Inhibitor Cocktail III, Animal Free (For Mammalian Cells) is a mixture of six protease inhibitors for use in applications that require animal free reagents.
Brand: Fisher BioReagents™ 12841640
Code : 32
Additional Details : Weight : 0.10000kg
Description
- Will treat proteases extracted from 20g tissue
- Specificity: Serine Proteases, Cyteine Proteases, Trypsin like Proteases, Aspartic Proteases, Aminapeptidases
Specifications
Protease Inhibitor Cocktail | |
Liquid | |
1 Vial | |
AEBSF HCl , Aprotinin (recombinant), Bestatin, E-64, Leupeptin, Pepstatin A |