Learn More
Trajan Scientific and Medical™ ProteCo™ PEEK Stainless Steel Unions
Precision engineered for capillary LC. Trajan Scientific and Medical ProteCol™ PEEK Stainless Steel Unions offer complete versatility for connecting capillary tubing.
Brand: Trajan Scientific and Medical™ 222006
Code : NEW
Additional Details : Weight : 0.01000kg
Description
- Leak free connections
- Replacement parts such as nuts and ferrules
Specifications
Union | |
Size: 1/32 in. to 0.36 mm | |
Stainless Steel |