missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ Protection Filter Set
Designed for use with Eppendorf Reference™ 2, Eppendorf Research™ plus and Eppendorf Xplorer™ (plus) pipettes
Brand: Eppendorf™ 4920623005
Code : NEW
Additional Details : Weight : 0.01100kg
Description
- Available in a variety of colors
- Protection Filter Sets include 10 filters and 1 filter sleeve
- For fixed- and variable-volume pipettes
Specifications
Violet | |
5 mL | |
10 filters with 1 filter sleeve |
Protection filter | |
0.5 to 5mL variable Eppendorf Reference™ 2, Eppendorf Research plus and Eppendorf Xplorer™ (plus) | |
1 Each |