Learn More
3M™ Protective Coverall 4520CS
81.60 EUR valid until 2024-12-31
Use promo code "22912" to get your promotional price.
Use promo code "22912" to get your promotional price.
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
Description
- Breathable material technology for reduced heat stress and premium wearer comfort all day
- Made from super breathable SMMMS non-woven for effective release of body heat and moisture and reduced heat stress
- Standard collar for use with respiratory equipment or where a hood may not be desired
- Knitted cuffs and elasticated waist and ankles for increased comfort and freedom of movement
- Anti-static coating EN 1149-5 reduces static discharge and the risk of sparks
- EN 1073-2 nuclear particulate hold out
- Knitted cuffs for soft and secure arm placement
Specifications
Product Type | Protective Coveralls |
Size | 2X-Large |
Material | SMS Fabric |
Color | White |
Waist Style | Elastic |
Cuff Style | Knit |
Closure Type | 2-way Zipper |
Seam Type | Serged |
Certifications/Compliance | CE Category III, EN 1073-2 Particulate Radioactive Contamination, EN 1149-5 - Electrostatic Properties, EN 13034:2005+A1:2009 Type 6 Liquid Splash, EN ISO 13982-1:2004 Type 5 Hazardous Dusts |