Learn More
Thermo Scientific™ Proteinase K, Tritirachium album limber
Commonly used in molecular biology to digest protein and remove contamination from preparations of nucleic acid. Addition of Proteinase K to nucleic acid preparations rapidly inactivates nucleases that might otherwise degrade the DNA or RNA during purification.
Brand:  Thermo Scientific™ J62051.MB
Code : NEW
Additional Details : CAS Number : 39450-01-6 Weight : 0.00025kg
 View more versions of this product
Product Code. 15474509
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
Keep cold
25 mg
Lyophilized Powder
Product Suggestions
Product Suggestions
Videos
SDS Documents