missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSAP Mouse, Clone: SPM312, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 55-MSM3X-P1
Additional Details : Weight : 0.01000kg
Specifications
PSAP | |
Cytoplasmic | |
SPM312 | |
Unconjugated | |
NP_001127666 | |
Mouse | |
52kDa | |
Protein A/G Purified | |
RUO | |
55 | |
IgG1 κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
ACPP | |
Prostatic acid phosphatase purified from human seminal plasma | |
PC12 cells or normal prostate or prostate carcinoma | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |