missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSF™ Terrain Safety Trainer Shoes
42.59 € - 54.90 €
Specifications
Color | Black |
---|---|
Material | Leather, Mesh |
Sole Features | Dual Density |
Sole Material | Polyurethane |
Style | Trainer |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
12802296
|
PSF™
980NMP-9 |
9 |
54.90 €
1 pair |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||||
12882286
|
PSF™
980NMP-7 |
7 |
54.90 €
1 pair |
Estimated Shipment: 05-07-2024
Log in to see stock. |
|||||
12822286
|
PSF™
980NMP-12 |
12 |
54.90 €
1 pair |
Estimated Shipment: 05-07-2024
Log in to see stock. |
|||||
12872286
|
PSF™
980NMP-6 |
6 |
54.90 €
1 pair |
Estimated Shipment: 04-07-2024
Log in to see stock. |
|||||
12812286
|
PSF™
980NMP-11 |
11 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
Black | |
Dual Density | |
Trainer | |
Safety Trainer Shoe |
Leather, Mesh | |
Polyurethane | |
Heat, Water-resistant |