missing translation for 'onlineSavingsMsg'
Learn More

Kinematica™ Polytron™ 1200E, 1300D and 2500E Accessory Generator Probes

Polytron™ dispersing aggregates are available in different designs, like rotor with knives for pre-cutting, rotor/stator with several teeth rows for fine or double emulsions, mechanical seals for vacuum and pressure applications and more.

Brand:  Kinematica™ 11030030

Code : 04

Additional Details : Weight : 0.24000kg

 View more versions of this product

Product Code. 11955418

  • 613.00 € / Pack of 25


Estimated Shipment: 23-07-2021
Add to basket

Description

Description

Polytron™ dispersing aggregates are available in different designs, like rotor with knives for pre-cutting, rotor/stator with several teeth rows for fine or double emulsions, mechanical seals for vacuum and pressure applications and more.

  • Autoclave safe
  • Includes Instruction Manual and tool for easy disassembly
  • Click and Go Coupling System allows for one hand assembly
  • Warranty: 2 years
Specifications

Specifications

Diposable Sawtooth
25 Pk.
Dispersion, Emulsions, Size Reduction, Suspension, Deagglomerate, Encapsulations, Extractions
7 mm
Ideal for High Throughput Applications and to Further Avoid Cross Contamination
Polyoxymethylene
0.3-10mL
82mm
0.3 to 10 mL
82 mm
Probe
Polytron™ 1200 E, 1300 D, 2500 E Homogenizers
PT-DA 07/2 SYN-E082
6.5 m/s
Videos
SDS
Documents
Certificates