missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ PTFE Containers

Product Code. 10393882 Shop All Fisherbrand Products
Click to view available options
Capacity (Metric):
15 mL
30 mL
60 mL
120 mL
240 mL
360 mL
480 mL
1000 mL
2200 mL
Unit Size:
Each

Product Code. 10393882

Only null left
Add to basket
Add to basket

Isostatically molded from pure PTFE. Fisherbrand™ PTFE Containers contain thick wall construction.

  • Smooth internal finish
  • Contains crew cap
  • Stackable
  • Autoclavable

Specifications

Capacity (Metric) 480 mL
Capacity (English) 16 oz.
Color White
Diameter (Metric) Outer Neck 76mm
Diameter (Metric) Inner Neck 73mm
For Use With (Application) Storage and Transport of Hazardous Chemicals
Includes Closure
Height (Metric) With Closure 126mm
Material PTFE
Max. Temperature (Metric) 280°C
Temperature Range (Metric) -200 to +280°C
Thickness (Metric) 13 mm
Transparency No
Weight (Metric) 350 g
Graduations No
Certifications/Compliance Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
Closure Included Yes
Graduated No
Height (Metric) 125 mm
Product Type Storage Container
Closure Size 90 mm
Diameter (Metric) 90 mm
Closure Type Screw Cap
Autoclavable Autoclavable
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.