Learn More
Fisherbrand™ PTFE Joint Adapter, Full Length A Type
Fisherbrand™ PTFE Joint Adapters are grease-free, high vacuum seals for standard taper ground glass joints, with substantial shoulder for ease of handling and repeated use. High vacuum performance is attained using transverse sealing rings.
9.75 € - 32.45 €
Specifications
Description | Fisherbrand™ PTFE Joint Adapter, Full Length A Type |
---|---|
For Use With (Equipment) | Standard Taper Ground Glass Joints |
Material | PTFE |
Quantity | 1 |
Product Code | Brand | Standard Taper Joint No. | Cone Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Standard Taper Joint No. | Cone Size | Price | Quantity & Availability | ||||
16216011
|
Fisherbrand™
|
10/30 | 14/35 |
9.75 €
Each |
Estimated Shipment: 05-07-2024
Log in to see stock. |
||||
16226011
|
Fisherbrand™
|
10/30 | 19/38 |
10.70 €
Each |
Estimated Shipment: 05-07-2024
Log in to see stock. |
||||
16236011
|
Fisherbrand™
|
14/35 | 19/38 |
12.75 €
Each |
Estimated Shipment: 05-07-2024
Log in to see stock. |
||||
16246011
|
Fisherbrand™
|
14/35 | 24/40 |
15.75 €
Each |
Estimated Shipment: 05-07-2024
Log in to see stock. |
||||
16256011
|
Fisherbrand™
|
19/38 | 24/40 |
15.75 €
Each |
Estimated Shipment: 07-05-2024
Log in to see stock. |
||||
16266011
|
Fisherbrand™
|
19/38 | 29/42 |
18.80 €
Each |
Estimated Shipment: 05-07-2024
Log in to see stock. |
||||
16276011
|
Fisherbrand™
|
19/38 | 34/45 |
24.55 €
Each |
Estimated Shipment: 05-07-2024
Log in to see stock. |
||||
16286011
|
Fisherbrand™
|
24/40 | 29/42 |
22.30 €
Each |
Estimated Shipment: 07-05-2024
Log in to see stock. |
||||
16296011
|
Fisherbrand™
|
24/40 | 34/45 |
24.45 €
Each |
Estimated Shipment: 05-07-2024
Log in to see stock. |
||||
16206021
|
Fisherbrand™
|
24/40 | 45/50 |
32.45 €
Each |
Estimated Shipment: 05-07-2024
Log in to see stock. |
||||
16216021
|
Fisherbrand™
|
29/42 | 34/45 |
24.45 €
Each |
Estimated Shipment: 05-07-2024
Log in to see stock. |
||||
Description
- Suitable for high vacuum through the use of transverse trapezoidal sealing rings on the contacting surfaces
- Use up to 280°C
- Eliminates the need for grease
- Leakage Rate: <1 x 10-4 torr, litre, sec-1
- Sleeves are inert
Specifications
Fisherbrand™ PTFE Joint Adapter, Full Length A Type | |
Standard Taper Ground Glass Joints | |
PTFE | |
1 |