missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bohlender™ BOLA™ PTFE Plugs for Screw Caps
Designed for safe plugging of unused ports of glass devices or GL connecting parts. PTFE plugs feature universal chemical resistance.
Brand: Bohlender™ D549-25
Additional Details : Weight : 0.50000kg
Description
- Plugs completely made of PTFE
- Suitable for replacement caps made of ETFE and PPS
- Plug is inserted into the cap and snaps in as soon as the cap is tightened
- Easily be removed for cleaning
Specifications
Plug | |
PTFE | |
-200°C | |
GL 25 |
FDA | |
For screw caps | |
1 Each | |
250°C |