Learn More
Poulten Graf™ Fortuna™ Optifix™ Basic Dispenser
Poulten Graf™ Fortuna™ Optifix™ Basic Dispensers have a dispensing cylinder and piston made of glass.
Brand:  Poulten Graf™ 101 08038
Code : 03
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10518151
  • 415.00 € / Each
Estimated Shipment: 19-06-2024
to see stock.
Add to basket
Description
Description
For dosing of the most common chemicals, acids and crytallizing liquids (not suitable for solvents). Poulten Graf™ PTFE/PP/Fiberglass/Teflon/Ceramic Fortuna™ Optifix™ Basic Dispensers have a dispensing cylinder and piston made of glass.
  • Manufactured according to DIN EN ISO 9001:2008 using high quality materials
  • With solid PTFE encased dispensing piston made of glass
  • Cylinder coated with plastic to protect against breakage
  • With certificate, individually numbered
  • Ergonomic finger protection cap made of PP with volume setting system
  • Graduation on the scale is lasered and easy to read
  • Fast and easy volume setting with additional fine adjustment
Specifications
Specifications
Fortuna Optifix Basic Dispenser
PTFE, Polypropylene, Fiberglass, Teflon, Ceramic
For dispensing aqueous solutions, acids and crystallizing liquids
Clear
2 to 10 mL