Learn More
BRAND™ PTFE Watch Glass
PTFE construction. Ideal for covering PTFE beakers.
Brand:  BRAND™ 150902
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 10415651
  • 8.55 € / Each
Estimated Shipment: 26-06-2024
to see stock.
Add to basket
Description
Description
  • PTFE construction
  • Excellent chemical resistance
  • Ideal for covering PTFE beakers
Specifications
Specifications
Flat
Round
30 mm
PTFE
Watch Glass