missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pumice Stone, Honeywell Fluka™
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Poly Bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Color | Gray |
Physical Form | Granulated |
Quantity | 500 g |
Packaging | Poly Bottle |
UN Number | NONH for all modes of transport |
Chemical Name or Material | Pumice Stone |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'