Learn More
Welch Ilmvac™ Pump P23Z vacuum, rotary vane 230V 50/60Hz 522mm x 171mm x 256mm ultimate vacuum 1 x 10-3 pumping speed 21.0m3/hr Ilmvac
Brand:  Welch Ilmvac™ 322007
Code : 50
Additional Details : Weight : 50.00000kg
Product Code. 12682885