Learn More
Thermo Scientific Chemicals Puromycin dihydrochloride hydrate, 99%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Information
• Puromycin dihydrochloride hydrate is the hydrochloride salt form of puromycin and is an aminoglycoside with antibiotic activity isolated from the Streptomyces alboniger
• Puromycin dihydrochloride hydrate is a protein synthesis inhibitor that causes premature chain termination by acting as an analog of the 3’-terminal ends of aminoacyl-tRNA, inhibiting protein synthesis
Applications
• This compound shows anti-microbial, anti-protozoal, anti-trypanosomal, and anti-neoplastic activities and is also used as an antibiotic in cell culture
• It also reveals anti-tumor activity in studies on brain cancer cells
• This compound has been used to study transcriptional regulatory mechanisms that control the sequential and coordinate expression of genes during cell differentiation
• It is active against gram-positive microorganisms, less active against acid-fast bacilli and gram-negative microorganisms. Puromycin can prevent growth of bacteria, protozoa, algae, and mammalian cells in in vitro studies.
Specifications
Melting Point | 168.0°C to 178.0°C |
Assay Percent Range | 98.5% min. (HPLC) |
Packaging | Glass bottle |
Quantity | 10 mg |
Beilstein | 26,IV,3704 |
Merck Index | 15,8054 |
Solubility Information | Solubility in water: soluble. Other solubilities: less soluble in ethanol |
Formula Weight | 544.44 |
Percent Purity | 99% |
Chemical Name or Material | Puromycin dihydrochloride hydrate |
Safety and Handling
- Puromycin dihydrochloride hydrate
- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
RUO – Research Use Only