missing translation for 'onlineSavingsMsg'
Learn More

PURUS™ 60-Layer Biohybrid&trade Contamination Control Mats

PURUS&trade Biohybrid&trade Mats are the first ever contamination control products that contain renewable plant-based materials, providing an environmentally friendly alternative to removing dirt and dust from foot traffic and equipment wheels

Brand:  PURUS™ PM 3645 64 W-HT

Code : NEW

Additional Details : Weight : 8.70000kg

 View more versions of this product

Product Code. 15391992

  • 262.00 € / Pack of 4


Estimated Shipment
13-06-2019
Add to basket

Description

Description

  • Earth Friendly containing GMO free, renewable plant based Cardia Biohybrid&trade material that reduces CO2 emissions
  • Strong & Effective delivering reliable and high-end performance
  • Consistent adhesive coverage for maximum performance of each layer
  • Can be recycled in conventional plastic recycling streams
  • Low profile numbered tabs ensure one-sheet-at-a-time removal and indicate number of remaining layers

Certifications

Manufactured in ISO 9001 certified facility

Specifications

Specifications

LDPE Blend
36 x 45 in. (91.4 x 114.3 cm)
Layer Thickness: 1.4 mil
White
For use in Pharmaceutical, Biotech, Semiconductor, Cleanrooms, and Industrial Applications
Videos