missing translation for 'onlineSavingsMsg'
Learn More

PURUS™ 60-Layer EnviroTack™ Contamination Control Mats

Exclusively from PURUS™, EnviroTack™ mats are the first ever contamination control products that contain GMO free, plant-based renewable materials, providing an environmentally friendly alternative to removing dirt and dust from foot traffic and equipment wheels

Brand:  PURUS™ PM 1836 64 G

Code : NEW

Additional Details : Weight : 3.60000kg

 View more versions of this product

Product Code. 15321962

  • 153.00 € / Pack of 4


Estimated Shipment
15-07-2020
Add to basket

Description

Description

  • Eco-friendly containing a proprietary GMO free, plant based renewable material that reduces CO2 emissions during the manufacturing process
  • Strong & Effective delivering reliable and high-end performance
  • Consistent adhesive coverage for maximum performance of each layer
  • Can be recycled in conventional plastic recycling streams or through Fisher Scientific's partnership with PURUS™ and Terra Cycle
  • Low profile numbered tabs ensure one-sheet-at-a-time removal and indicate number of remaining layers

Certifications

Manufactured in ISO 9001 certified facility

Specifications

Specifications

Manufactured in an ISO 9001 certified facility
60 layers per mat, 4 mats per case
LDPE Blend
457 x 914 mm
Film Thickness: 1.3 ± 0.2 mil / 33 μm Adhesive Thickness: 0.3 mil/ 8 μm
Safety
Regular Tack
Gray
For use in Pharmaceutical, Biotech, Semiconductor, Cleanrooms, and Industrial Applications
Videos
Documents