missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ PVT1 Split FISH Probe
Labeled FISH probes for identification of gene split using Fluoresecent In Situ Hybridization Technique.
1475.00 € - 2430.00 €
Specifications
Content And Storage | Store at 4°C in the dark |
---|---|
Format | Tube |
For Use With (Application) | Fluorescent In Situ Hybridization (Cell) |
Gene | Pvt1 oncogene (non-protein coding) |
Gene Alias | MGC21751,NCRNA00079 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16023495
|
Abnova™
FS0053.100UL |
100 μL |
1475.00 €
100µL |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16033495
|
Abnova™
FS0053.200UL |
200 μL |
2430.00 €
200µL |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Store at 4°C in the dark | |
Fluorescent In Situ Hybridization (Cell) | |
MGC21751,NCRNA00079 | |
5820 | |
Liquid |
Tube | |
Pvt1 oncogene (non-protein coding) | |
PVT1 | |
DNA/RNA |