missing translation for 'onlineSavingsMsg'
Learn More

Pyrazinecarbonitrile, 99%, ACROS Organics™

Brand:  Acros Organics 436920100

Code : 29

Additional Details : CAS Number : 19847-12-2 Weight : 0.06000kg Transport : UN number : 3276 Chem class : 6.1 Pack group : III


Packaging Glass bottle
Quantity 10g

 View more versions of this product

Product Code. 11946651

  • 17.48 € / 10g

In stock, ready to ship: 1

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

19847-12-2
105.10
PMSVVUSIPKHUMT-UHFFFAOYSA-N
73172
pyrazine-2-carbonitrile
C5H3N3
MFCD00049361
pyrazinecarbonitrile, 2-cyanopyrazine, cyanopyrazine, 2-pyrazinecarbonitrile, pyrazinenitrile, pyrazinonitrile, 2-cyanopyarine, 2-cyano pyrazine
CHEBI:3982
N#CC1=CN=CC=N1

Specifications

Pyrazinecarbonitrile
97°C
Glass bottle
1.174
Colorless to Yellow
10g
98.5% min. (GC)
MFCD00049361
Solubility in water: soluble
N#CC1=CN=CC=N1
105.10
CHEBI:3982
Liquid
1.1740g/mL
Authentic
1.5330 to 1.5350 (20°C, 589nm)
90°C to 93°C (15.0mmHg)
20°C
19847-12-2
C5H3N3
pyrazinecarbonitrile, 2-cyanopyrazine, cyanopyrazine, 2-pyrazinecarbonitrile, pyrazinenitrile, pyrazinonitrile, 2-cyanopyarine, 2-cyano pyrazine
PMSVVUSIPKHUMT-UHFFFAOYSA-N
pyrazine-2-carbonitrile
73172
105.1
99%
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2-Pyrazinecarbonitrile

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS Documents
Certificates