missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyrex™ 30mm Diameter Glass Büchner Filter Funnel
Manufactured from chemically resistant Pyrex™ borosilicate glass. Pyrex™ 30mm Glass Büchner Filter Funnnel includes an integral sintered glass disc also constructed with Pyrex™ glass. High resistance to most corrosive agents, and does not require filter papers.
203.00 € - 840.00 €
Specifications
Capacity (Metric) | 35 mL |
---|---|
Diameter (Metric) Stem | 8mm |
Material | Pyrex Glass, Borosilicate Glass |
Height (Metric) | 160 mm |
For Use With (Application) | For convenient filtration of solutions |
Product Code | Brand | Porosity | Pore Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Porosity | Pore Size | Price | Quantity & Availability | ||||
11971025
|
Pyrex
3620/06 |
3 | 16 to 40μm |
271.00 €
Pack of 5 |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||||
12633212
|
Pyrex
3620/02 |
1 | 100 to 160μm |
203.00 €
Pack of 5 |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||||
12643212
|
Pyrex
3620/04 |
2 | 40 to 100μm |
279.00 €
Pack of 5 |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||||
11981548
|
Pyrex
3620/08 |
4 | 10 to 16μm |
203.00 €
Pack of 5 |
Estimated Shipment: 21-08-2024
Log in to see stock. |
||||
15537710
|
Pyrex
3620/10 |
5 | 1.0 to 1.6μm |
840.00 €
Pack of 20 |
Estimated Shipment: 08-10-2024
Log in to see stock. |
||||
Description
Manufactured from chemically resistant Pyrex™ borosilicate glass. Pyrex™ 30mm Glass Büchner Filter Funnnel includes an integral sintered glass disc also constructed with Pyrex™ glass. High resistance to most corrosive agents, and does not require filter papers.
- Comprehensive range of Büchner filter funnels with integral sintered glass discs
- Disc and funnel body manufactured from chemically resistant Pyrex™ glass
- For convenient filtration of solutions - No filter papers required - Highly resistant to most corrosive reagents including ammonia and concentrated sulfuric acid
- Choice of disc diameters and porosities
Specifications
35 mL | |
Pyrex Glass, Borosilicate Glass | |
For convenient filtration of solutions | |
Integrated filter plate sintered Glass disc | |
Buchner filter funnel |
8mm | |
160 mm | |
30mm | |
100mm |